Assay Details
Target Gene Details
Entrez Gene ID: | 81877 |
Gene Name: | tenascin XB |
Gene Aliases: |
Tn-mhc, Tnx |
Location: |
Chr.17:34659788-34795460 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnxb | NM_031176.2 | NP_112453.2 | ||
| AB235155.1 | BAE44473.1 | |||
| AK136535.1 | ||||
| AK171696.1 | BAE42616.1 | |||
| X73959.1 | CAA52147.1 |
More Information
Set Membership: |
|

Genomic Map