Assay Details
Target Gene Details
Entrez Gene ID: | 110695 |
Gene Name: | aldehyde dehydrogenase family 7, member A1 |
Gene Aliases: |
Atq1, D18Wsu181e |
Location: |
Chr.18:56524722-56572939 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh7a1 | NM_001127338.1 | NP_001120810.1 | ||
| AK004991.1 | BAB23726.1 | |||
| CJ156580.1 |
Target Gene Details
Entrez Gene ID: | 56698 |
Gene Name: | phosphorylated adaptor for RNA export |
Gene Aliases: |
2810055C14Rik, 4933427L19Rik, AU018701, AU018854, D18Ertd65e, Rnuxa, p55 |
Location: |
Chr.18:56562569-56587712 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phax | NM_001162989.1 | NP_001156461.1 | ||
| AK016958.1 |
More Information
Set Membership: |
|

Genomic Map