Assay Details
Target Gene Details
Entrez Gene ID: | 16948 |
Gene Name: | lysyl oxidase |
Gene Aliases: |
AI893619, TSC-160, rrg |
Location: |
Chr.18:52516060-52529867 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lox | NM_001286181.1 | NP_001273110.1 | ||
| NM_001286182.1 | NP_001273111.1 | |||
| NM_010728.3 | NP_034858.2 | |||
| AK135213.1 | BAE22461.1 | |||
| AK141995.1 | BAE24907.1 | |||
| AK159199.1 | ||||
| AK159241.1 | BAE34923.1 | |||
| AK159263.1 | BAE34943.1 | |||
| AK159368.1 | BAE35026.1 | |||
| AK159682.1 | BAE35285.1 | |||
| AK159796.1 | BAE35377.1 | |||
| AK160096.1 | BAE35626.1 | |||
| AK164634.1 | BAE37854.1 | |||
| AK166279.1 | BAE38676.1 | |||
| BC018439.1 | AAH18439.1 | |||
| M65142.1 | AAA19032.1 | |||
| M65143.1 | AAA20185.1 |
More Information
Set Membership: |
|

Genomic Map