Assay Details
Target Gene Details
Entrez Gene ID: | 69256 |
Gene Name: | zinc finger protein 397 |
Gene Aliases: |
2810411K16Rik, 6720480F11Rik, AI461664 |
Location: |
Chr.18:23954662-23964671 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp397 | NM_027007.2 | NP_081283.2 | ||
| AK013086.1 | BAB28640.1 | |||
| AK032001.1 | BAC27643.1 | |||
| AK078454.1 | 1 | 2388 | ||
| AY338468.1 | AAQ15292.1 |
More Information
Set Membership: |
|

Genomic Map