Assay Details
Target Gene Details
Entrez Gene ID: | 56248 |
Gene Name: | adenylate kinase 3 |
Gene Aliases: |
AA407498, AI506714, AK-3, Ak3l, Ak3l1, Akl3l |
Location: |
Chr.19:28995380-29048672 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ak3 | NM_021299.1 | 4 | 574 | NP_067274.1 |
| AB020203.1 | 4 | 477 | BAA77360.1 | |
| AK004864.2 | 4 | 589 | BAB23625.1 | |
| AK005194.2 | 4 | 628 | BAB23876.1 | |
| AK008681.1 | 4 | 591 | BAB25829.1 | |
| AK031631.1 | 4 | 572 | BAC27488.1 | |
| AK053644.1 | 4 | 634 | BAC35459.1 | |
| AK089027.1 | 4 | 625 | BAC40707.1 | |
| AK147060.1 | 4 | 634 | BAE27644.1 | |
| AK149996.1 | 4 | 631 | BAE29224.1 | |
| BC016432.1 | 4 | 576 | AAH16432.1 | |
| BC019174.1 | 4 | 627 | AAH19174.1 | |
| BC024871.1 | 4 | 574 | AAH24871.1 | |
| BC058191.1 | 4 | 604 | AAH58191.1 |
More Information
Set Membership: |
|

Genomic Map