Assay Details
Target Gene Details
Entrez Gene ID: | 11668 |
Gene Name: | aldehyde dehydrogenase family 1, subfamily A1 |
Gene Aliases: |
Ahd-2, Ahd2, Aldh1, Aldh1a2, E1, Raldh1 |
Location: |
Chr.19:20601982-20643462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh1a1 | NM_013467.3 | NP_038495.2 | ||
| BC044729.2 | AAH44729.1 | |||
| BC054386.1 | AAH54386.1 | |||
| M74570.1 | AAA37202.1 | |||
| S75713.1 | AAB32754.2 |
More Information
Set Membership: |
|

Genomic Map