Assay Details
Target Gene Details
Entrez Gene ID: | 67180 |
Gene Name: | Yip1 domain family, member 5 |
Gene Aliases: |
2610311I19Rik, AA408236, Yip1a |
Location: |
Chr.18:40204865-40219399 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yipf5 | NM_023311.3 | NP_075800.1 | ||
| AF140226.1 | AAG48522.1 | |||
| AK007671.1 | ||||
| AK010089.1 | BAB26694.1 | |||
| AK151425.1 | BAE30389.1 | |||
| AK151461.1 | BAE30419.1 | |||
| AK152440.1 | BAE31221.1 | |||
| AK153123.1 | BAE31738.1 | |||
| AK160004.1 | BAE35552.1 | |||
| AK160270.1 | ||||
| AK167526.1 | BAE39596.1 | |||
| AK167953.1 | ||||
| AK172133.1 | ||||
| AK181274.1 | ||||
| AK205561.1 | ||||
| AK207818.1 | ||||
| AK212051.1 | ||||
| AK218592.1 | ||||
| BC003317.1 | AAH03317.1 | |||
| BC055301.1 | AAH55301.1 |

Genomic Map