Assay Details
Target Gene Details
Entrez Gene ID: | 75138 |
Gene Name: | RIKEN cDNA 4930526L06 gene |
Gene Aliases: |
- |
Location: |
Chr.19:11196817-11299884 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930526L06Rik | NR_045317.1 | |||
| AK015905.1 |
Target Gene Details
Entrez Gene ID: | 269063 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 5 |
Gene Aliases: |
- |
Location: |
Chr.19:11273866-11283895 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a5 | NM_183190.2 | NP_899013.1 | ||
| AF544401.1 | AAQ11743.1 | |||
| AY039035.1 | AAK92695.1 | |||
| AY039036.1 | AAK92696.1 | |||
| BC049657.1 | AAH49657.1 |
More Information
Set Membership: |
|

Genomic Map