Assay Details
Target Gene Details
Entrez Gene ID: | 69745 |
Gene Name: | polymerase (DNA-directed), delta 4 |
Gene Aliases: |
2410012M21Rik, AI463381, AW060307, Polds, p12 |
Location: |
Chr.19:4231899-4233634 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pold4 | NM_027196.3 | NP_081472.1 | ||
| AF515709.1 | AAM53977.1 | |||
| AK010477.1 | BAB26970.1 | |||
| AK170469.1 | ||||
| BC028520.1 | AAH28520.1 |
More Information
Set Membership: |
|

Genomic Map