Assay Details
Target Gene Details
Entrez Gene ID: | 75138 |
Gene Name: | RIKEN cDNA 4930526L06 gene |
Gene Aliases: |
- |
Location: |
Chr.19:11196817-11299884 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930526L06Rik | NR_045317.1 | |||
| AK015905.1 |
Target Gene Details
Entrez Gene ID: | 12482 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 1 |
Gene Aliases: |
AA960661, Cd20, Ly-44, Ms4a2 |
Location: |
Chr.19:11250603-11266151 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a1 | NM_007641.5 | NP_031667.1 | ||
| AK017903.1 | BAB30996.1 | |||
| AK041428.1 | BAC30944.1 | |||
| AK080040.1 | BAC37812.1 | |||
| AK137469.1 | ||||
| AK143824.1 | ||||
| AK166571.1 | 1 | 70 | ||
| AK166659.1 | 1 | 70 | ||
| AK171631.1 | BAE42576.1 | |||
| AK171659.1 | BAE42593.1 | |||
| BC028322.1 | AAH28322.1 |
More Information
Set Membership: |
|

Genomic Map