Assay Details
Target Gene Details
Entrez Gene ID: | 192176 |
Gene Name: | filamin, alpha |
Gene Aliases: |
ABP-280, Dilp2, F730004A14Rik, Fln1, GENA 379, filamin-1 |
Location: |
Chr.X:74223461-74249854 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 33 - Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flna | NM_001290421.1 | NP_001277350.1 | ||
| NM_010227.3 | NP_034357.2 | |||
| AK131179.1 | BAD21429.1 | |||
| BC151024.1 |

Genomic Map