Assay Details
Target Gene Details
Entrez Gene ID: | 100039138 |
Gene Name: | predicted gene 2061 |
Gene Aliases: |
- |
Location: |
Chr.1:185447116-185455568 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm2061 | NR_040305.1 | 1 | 169 | |
| AK138296.1 | 1 | 170 |
Target Gene Details
Entrez Gene ID: | 226781 |
Gene Name: | solute carrier family 30, member 10 |
Gene Aliases: |
E130106K10Rik, Gm212 |
Location: |
Chr.1:185454848-185468762 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc30a10 | NM_001033286.2 | 1 | 553 | NP_001028458.1 |
| AK053537.1 | 1 | 554 | ||
| AK136932.1 | 1 | 361 | BAE23176.1 | |
| BB665136.1 | 1 | 554 |

Genomic Map