Assay Details
Target Gene Details
Entrez Gene ID: | 12765 |
Gene Name: | chemokine (C-X-C motif) receptor 2 |
Gene Aliases: |
CD128, CDw128, Cmkar2, Gpcr16, IL-8Rh, IL-8rb, IL8RA, Il8rb, mIL-8RH |
Location: |
Chr.1:74153994-74161246 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxcr2 | NM_009909.3 | 2 | 614 | NP_034039.1 |
| AK158234.1 | 3 | 620 | BAE34417.1 | |
| CJ056061.1 | 2 | 615 | ||
| D17630.1 | 2 | 610 | BAA04536.1 | |
| L20337.1 | AAA16853.1 |

Genomic Map