Assay Details
Target Gene Details
Entrez Gene ID: | 620246 |
Gene Name: | G protein-coupled receptor 52 |
Gene Aliases: |
EG620246 |
Location: |
Chr.1:160576668-160577753 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr52 | NM_001146330.1 | 1 | 546 | NP_001139802.1 |
Target Gene Details
Entrez Gene ID: | 29809 |
Gene Name: | RAB GTPase activating protein 1-like |
Gene Aliases: |
5830411O09Rik, 8430421H08Rik, 9630005B12Rik, AW049894, HHL, Hh1, mKIAA0471 |
Location: |
Chr.1:160219174-160793476 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabgap1l | NM_013862.5 | NP_038890.3 | ||
| AK035796.1 | BAC29189.1 | |||
| BC145811.1 | ||||
| BC145813.1 |
More Information
Set Membership: |
|

Genomic Map