Assay Details
Target Gene Details
Entrez Gene ID: | 228836 |
Gene Name: | discs, large homolog-associated protein 4 (Drosophila) |
Gene Aliases: |
AI225853, BC024558, DAP-4, DAP4, SAPAP-4, Sapap4, WBP16 |
Location: |
Chr.2:156613633-156764363 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlgap4 | NM_001042487.1 | 2 | 1010 | NP_001035952.1 |
| NM_001042488.2 | 2 | 309 | NP_001035953.1 | |
| NM_001277186.1 | 8 | 2254 | NP_001264115.1 | |
| NM_001277187.1 | 7 | 2203 | NP_001264116.1 | |
| NM_146128.6 | 8 | 2254 | NP_666240.4 | |
| AK089484.1 | 2 | 230 | ||
| AK122408.1 | 8 | 2254 | BAC65690.2 | |
| AK150554.1 | 2 | 230 | BAE29656.1 | |
| AK156212.1 | 2 | 230 | ||
| AK211649.1 | 2 | 220 | ||
| AY243849.2 | 6 | 1834 | AAO89220.2 | |
| BC024558.1 | 2 | 414 | AAH24558.1 | |
| BC058948.1 | 2 | 333 | AAH58948.1 | |
| BC085475.1 | 2 | 797 | AAH85475.1 | |
| BC106094.1 | 2 | 392 | AAI06095.1 | |
| BC141110.1 | 6 | 1891 | ||
| BC145394.1 | 6 | 1891 | ||
| BC145395.1 | 5 | 1840 | ||
| BY298512.1 | 2 | 290 |
More Information
Set Membership: |
|

Genomic Map