Assay Details
Target Gene Details
Entrez Gene ID: | 228836 |
Gene Name: | discs, large homolog-associated protein 4 (Drosophila) |
Gene Aliases: |
AI225853, BC024558, DAP-4, DAP4, SAPAP-4, Sapap4, WBP16 |
Location: |
Chr.2:156613633-156764363 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlgap4 | NM_001042487.1 | 4 | 1617 | NP_001035952.1 |
| NM_001042488.2 | 4 | 916 | NP_001035953.1 | |
| NM_001277186.1 | 10 | 2852 | NP_001264115.1 | |
| NM_001277187.1 | 9 | 2810 | NP_001264116.1 | |
| NM_146128.6 | 10 | 2861 | NP_666240.4 | |
| AK122408.1 | 8 | 5519 | BAC65690.2 | |
| AK150554.1 | 4 | 837 | BAE29656.1 | |
| AK156212.1 | 2 | 3495 | ||
| AK178050.1 | 1 | 41 | ||
| AK180513.1 | 1 | 41 | ||
| AK187414.1 | 1 | 41 | ||
| AY243849.2 | 8 | 2438 | AAO89220.2 | |
| BC024558.1 | 4 | 1021 | AAH24558.1 | |
| BC058948.1 | 4 | 940 | AAH58948.1 | |
| BC085475.1 | 4 | 1404 | AAH85475.1 | |
| BC106094.1 | 4 | 999 | AAI06095.1 | |
| BC141110.1 | 8 | 2489 | ||
| BC145394.1 | 8 | 2489 | ||
| BC145395.1 | 7 | 2447 |
More Information
Set Membership: |
|

Genomic Map