Assay Details
Target Gene Details
Entrez Gene ID: | 75823 |
Gene Name: | family with sequence similarity 227, member B |
Gene Aliases: |
4930525F21Rik |
Location: |
Chr.2:125983483-126152087 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam227b | NM_029455.3 | NP_083731.2 | ||
| AK015893.1 | BAB30022.1 |
Target Gene Details
Entrez Gene ID: | 14178 |
Gene Name: | fibroblast growth factor 7 |
Gene Aliases: |
Kgf |
Location: |
Chr.2:126001108-126091185 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf7 | NM_008008.4 | 4 | 2359 | NP_032034.1 |
| AK086639.1 | 5 | 2309 | BAC39707.1 |
More Information
Set Membership: |
|

Genomic Map