Assay Details
Target Gene Details
Entrez Gene ID: | 99151 |
Gene Name: | cerebral endothelial cell adhesion molecule |
Gene Aliases: |
2600006K01Rik, AL024097, Ceecam1 |
Location: |
Chr.2:29845105-29882845 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cercam |
