Assay Details
Target Gene Details
Entrez Gene ID: | 20269 |
Gene Name: | sodium channel, voltage-gated, type III, alpha |
Gene Aliases: |
Gm1000, Nav1.3 |
Location: |
Chr.2:65457118-65567527 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn3a | NM_018732.3 | 11 | 1850 | NP_061202.3 |
| AK148881.1 | 10 | 2559 | BAE28688.1 |
More Information
Set Membership: |
|

Genomic Map