Assay Details
Target Gene Details
Entrez Gene ID: | 69596 |
Gene Name: | adaptor-related protein 5 complex, sigma 1 subunit |
Gene Aliases: |
0610038L13Rik, 2310035K24Rik, AW060727, sigma5 |
Location: |
Chr.2:131210360-131213514 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap5s1 | NM_001291031.1 | 3 | 1143 | NP_001277960.1 |
| NM_001291032.1 | NP_001277961.1 | |||
| NM_027129.3 | 4 | 1054 | NP_081405.2 | |
| NR_110992.1 | 2 | 932 | ||
| AK002807.1 | 3 | 1068 | BAC25007.1 | |
| AK009634.1 | BAB26405.1 | |||
| AW060727.1 | ||||
| BC048399.1 | 4 | 974 | AAH48399.1 |

Genomic Map