Assay Details
Target Gene Details
Entrez Gene ID: | 12908 |
Gene Name: | carnitine acetyltransferase |
Gene Aliases: |
AW107812, CARAT |
Location: |
Chr.2:30400476-30415748 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crat | NM_007760.3 | 14 | 4008 | NP_031786.2 |
| AK028666.1 | 8 | 2651 | ||
| AK132179.1 | 15 | 3835 | BAE21016.1 |
More Information
Set Membership: |
|

Genomic Map