Assay Details
Target Gene Details
Entrez Gene ID: | 545477 |
Gene Name: | BPI fold containing family A, member 6 |
Gene Aliases: |
EG545477, Gm5840, Splunc6 |
Location: |
Chr.2:153974945-154000495 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifa6 | NM_001080811.1 | 4 | 586 | NP_001074280.1 |
| BC082608.1 | 4 | 586 |

Genomic Map