Assay Details
Target Gene Details
Entrez Gene ID: | 66617 |
Gene Name: | N-terminal Xaa-Pro-Lys N-methyltransferase 1 |
Gene Aliases: |
2610205E22Rik, AL033331, AL033332, Mettl11a, NTM1A |
Location: |
Chr.2:30807780-30823025 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntmt1 | NM_170592.2 | 4 | 992 | NP_733480.1 |
| AK009056.1 | 4 | 938 | ||
| AK011888.1 | 4 | 1002 | ||
| AK011960.1 | 4 | 994 | ||
| AK154783.1 | 4 | 1002 | BAE32826.1 | |
| AK179678.1 | ||||
| AK185556.1 | ||||
| AK188316.1 | ||||
| AK192866.1 | 1 | 261 | ||
| AK197307.1 | ||||
| AK205636.1 | ||||
| AK206716.1 | ||||
| AK207933.1 | ||||
| AK209177.1 | 1 | 162 | ||
| AK210956.1 | 1 | 141 | ||
| AK211177.1 | ||||
| AK215578.1 | ||||
| AK216805.1 | ||||
| AK218974.1 | ||||
| BC027220.1 | 4 | 992 | AAH27220.1 |
More Information
Set Membership: |
|

Genomic Map