Assay Details
Target Gene Details
Entrez Gene ID: | 71103 |
Gene Name: | glycosyltransferase 6 domain containing 1 |
Gene Aliases: |
4933411C14Rik, GT6m7 |
Location: |
Chr.2:25771820-25815940 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glt6d1 | NM_001039095.1 | 5 | 965 | NP_001034184.1 |
| AK016765.1 | 5 | 973 | ||
| BC111823.1 | 5 | 965 | AAI11824.1 |
More Information
Set Membership: |
|

Genomic Map