Assay Details
Target Gene Details
Entrez Gene ID: | 329540 |
Gene Name: | nucleolar protein 4-like |
Gene Aliases: |
4931410H15, 8430427H17Rik, AI429157, Gm1636 |
Location: |
Chr.2:153407461-153529971 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol4l | NM_001001986.2 | 3 | 501 | NP_001001986.2 |
| NM_001134300.2 | 6 | 1286 | NP_001127772.1 | |
| AK139459.1 | 3 | 501 | BAE24020.1 | |
| AK147524.1 | 6 | 1288 | BAE27972.1 | |
| AK157863.1 | 3 | 482 | BAE34240.1 | |
| BC075648.1 | 5 | 697 | AAH75648.1 | |
| BC079552.1 | 3 | 443 | AAH79552.1 | |
| BC094234.1 | 2 | 352 | AAH94234.1 |

Genomic Map