Assay Details
Target Gene Details
Entrez Gene ID: | 65112 |
Gene Name: | prostate transmembrane protein, androgen induced 1 |
Gene Aliases: |
2210418I02Rik, AW455466, Erg1.2, N4wbp4, Stag1, Tmepai |
Location: |
Chr.2:173224458-173276533 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmepa1 | NM_022995.3 | 4 | 3890 | NP_075371.2 |
| AK077382.1 | 1 | 2682 |

Genomic Map