Assay Details
Target Gene Details
Entrez Gene ID: | 100039684 |
Gene Name: | RIKEN cDNA 5031434O11 gene |
Gene Aliases: |
AI481280, Gm2370 |
Location: |
Chr.3:51559757-51567117 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5031434O11Rik | NR_033624.1 | 5 | 486 | |
| AI481280.1 | 1 | 234 | ||
| AK017144.1 | 4 | 1473 | ||
| AK048747.1 | 4 | 3498 | ||
| BB385113.2 | 3 | 426 | ||
| BX528204.1 | 5 | 487 |

Genomic Map