Assay Details
Target Gene Details
Entrez Gene ID: | 68527 |
Gene Name: | upper zone of growth plate and cartilage matrix associated |
Gene Aliases: |
1110017I16Rik, AW121955, Grp |
Location: |
Chr.2:4976122-4985748 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ucma | NM_001113558.2 | 5 | 595 | NP_001107030.1 |
| NM_001165932.1 | 4 | 496 | NP_001159404.1 | |
| NM_026754.3 | 4 | 529 | NP_081030.1 | |
| AK003750.1 | 4 | 471 | BAB22978.2 | |
| BC115608.2 | ||||
| BC115609.2 | ||||
| BY590633.1 | 1 | 171 | ||
| EF529510.1 | 5 | 566 | ||
| EU368184.1 | 5 | 595 | ||
| FJ217397.1 | 4 | 434 | ||
| FJ217398.1 | 3 | 368 | ||
| FJ217399.1 | 5 | 533 | ||
| FJ217400.1 | 4 | 467 |

Genomic Map