Assay Details
Target Gene Details
Entrez Gene ID: | 76089 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 2 |
Gene Aliases: |
5830453M24Rik, CNRasGEF, Pdzgef1, RA-GEF-1, mKIAA0313, nRapGEP |
Location: |
Chr.3:79062516-79287244 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapgef2 | NM_001099624.2 | 23 | 4260 | NP_001093094.2 |
| AB093228.1 | 16 | 3143 | BAC41412.1 | |
| AK147368.1 | 23 | 4188 | ||
| BC067056.1 | 23 | 4260 | AAH67056.1 |
More Information
Set Membership: |
|

Genomic Map