Assay Details
Target Gene Details
Entrez Gene ID: | 14745 |
Gene Name: | lysophosphatidic acid receptor 1 |
Gene Aliases: |
AI326300, Edg2, Gpcr26, Kdt2, lpA1, vzg-1 |
Location: |
Chr.4:58435252-58554027 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lpar1 | NM_001290486.1 | 3 | 2066 | NP_001277415.1 |
| NM_010336.2 | 4 | 2181 | NP_034466.2 | |
| NM_172989.1 | 4 | 2270 | NP_766577.1 | |
| AK030330.1 | 4 | 2234 | BAC26905.1 | |
| BC025425.1 | 4 | 2180 | AAH25425.1 | |
| U13370.1 | 1 | 500 | ||
| U70622.1 | AAC52923.1 |
More Information
Set Membership: |
|

Genomic Map