Assay Details
Target Gene Details
Entrez Gene ID: | 68364 |
Gene Name: | RIKEN cDNA 0610030E20 gene |
Gene Aliases: |
1500031N04Rik, 2810411C16Rik, AI461894 |
Location: |
Chr.6:72347023-72353162 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 0610030E20Rik | NM_026696.1 | 4 | 3246 | NP_080972.1 |
| BC066053.1 | 4 | 3220 |

Genomic Map