Assay Details
Target Gene Details
Entrez Gene ID: | 13401 |
Gene Name: | dystrophia myotonica-containing WD repeat motif |
Gene Aliases: |
DMR-N9, Dm9 |
Location: |
Chr.7:19076089-19082776 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmwd | NM_010058.2 | 1 | 163 | NP_034188.2 |

Genomic Map