Assay Details
Target Gene Details
Entrez Gene ID: | 69757 |
Gene Name: | leukocyte receptor cluster (LRC) member 1 |
Gene Aliases: |
1500001K17Rik |
Location: |
Chr.7:3660107-3665840 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Leng1 | NM_027203.3 | 1 | 91 | NP_081479.1 |
| BC023108.1 | 1 | 91 | AAH23108.1 |
Target Gene Details
Entrez Gene ID: | 353499 |
Gene Name: | transmembrane channel-like gene family 4 |
Gene Aliases: |
- |
Location: |
Chr.7:3665754-3677553 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmc4 | NM_181820.2 | NP_861541.2 | ||
| CN719678.1 |

Genomic Map