Assay Details
Target Gene Details
Entrez Gene ID: | 11933 |
Gene Name: | ATPase, Na+/K+ transporting, beta 3 polypeptide |
Gene Aliases: |
AA409958, AI664000, AW212096 |
Location: |
Chr.9:96332662-96364905 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp1b3 | NM_007502.4 | 7 | 1662 | NP_031528.1 |
| AK032516.1 | 7 | 1679 | BAC27906.1 | |
| AK151700.1 | 7 | 1679 | BAE30623.1 | |
| AK151841.1 | 7 | 1678 | BAE30733.1 | |
| AK159532.1 | 7 | 1679 | BAE35160.1 | |
| AK168273.1 | 7 | 1678 | BAE40219.1 | |
| AK178890.1 | 1 | 270 | ||
| AK181142.1 | ||||
| AK185963.1 | ||||
| AK205378.1 | 1 | 194 | ||
| AK206397.1 | 1 | 150 | ||
| AK213033.1 | 1 | 192 | ||
| AK214140.1 | ||||
| AK217181.1 | 1 | 234 | ||
| AK217555.1 | 1 | 177 | ||
| AK218295.1 | ||||
| AK218384.1 | ||||
| AK219621.1 | 1 | 201 | ||
| BC079916.1 | 7 | 1667 | ||
| U59761.1 | 7 | 1665 | AAC00019.1 |
More Information
Set Membership: |
|

Genomic Map