Assay Details
Target Gene Details
Entrez Gene ID: | 14479 |
Gene Name: | ubiquitin specific peptidase 15 |
Gene Aliases: |
4921514G19Rik, AI327321, E430033I05Rik, Gcap18 |
Location: |
Chr.10:123105006-123197019 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp15 | NM_001301628.1 | 15 | 2210 | NP_001288557.1 |
| NM_027604.4 | 16 | 2297 | NP_081880.2 | |
| AF468037.1 | 16 | 2201 | AAL77418.1 | |
| AK014891.1 | 10 | 1508 | ||
| AK122301.1 | 15 | 2125 | BAC65583.1 | |
| AK133852.1 | 15 | 2119 | BAE21887.1 | |
| AK145749.1 | 16 | 2201 | BAE26626.1 | |
| AK161469.1 | 16 | 2240 | BAE36413.1 | |
| AK161771.1 | 1 | 168 | BAE36566.1 | |
| AK161957.1 | 15 | 2158 | ||
| AK163315.1 | 15 | 2139 | ||
| AK168755.1 | 15 | 2145 | BAE40593.1 | |
| AK216706.1 | 2 | 300 | ||
| BC050042.1 | 16 | 2213 | AAH50042.1 |
More Information
Set Membership: |
|

Genomic Map