Assay Details
Target Gene Details
Entrez Gene ID: | 13998 |
Gene Name: | FYVE, RhoGEF and PH domain containing 6 |
Gene Aliases: |
AA123052, Etohd4, ZFYVE24 |
Location: |
Chr.10:94036001-94145339 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgd6 | NM_053072.3 | 21 | 7448 | NP_444302.4 |
| AK173157.1 | 21 | 7288 | BAD32435.1 |
More Information
Set Membership: |
|

Genomic Map