Assay Details
Target Gene Details
Entrez Gene ID: | 327992 |
Gene Name: | heat shock transcription factor family member 5 |
Gene Aliases: |
Gm739, MLZ-220 |
Location: |
Chr.11:87616404-87659881 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsf5 | NM_001045527.1 | 4 | 1396 | NP_001038992.1 |
More Information
Set Membership: |
|

Genomic Map