Assay Details
Target Gene Details
Entrez Gene ID: | 68585 |
Gene Name: | reticulon 4 |
Gene Aliases: |
1110020G17Rik, AA407876, AA409940, AA960376, ASY, C130026I10Rik, NOGO, NSP-CL, NgA, Nogo-A, Nogo-B, Nogo-C, mKIAA0886, mKIAA4153 |
Location: |
Chr.11:29692898-29744414 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtn4 | NM_024226.4 | 7 | 1148 | NP_077188.1 |
| NM_194051.3 | 9 | 3435 | NP_918940.1 | |
| NM_194052.3 | 7 | 1624 | NP_918941.1 | |
| NM_194053.3 | 8 | 1681 | NP_918942.1 | |
| NM_194054.3 | 9 | 4042 | NP_918943.1 | |
| AK003859.1 | 2 | 356 | ||
| AK034902.1 | 7 | 1568 | ||
| AK047253.1 | ||||
| AK186438.1 | ||||
| AK186869.1 | ||||
| AK213159.1 | 1 | 201 | ||
| AK218108.1 | 1 | 201 | ||
| AK220511.1 | 9 | 4002 | BAD90301.1 | |
| AY102280.1 | 9 | 3435 | AAM73502.1 | |
| AY102281.1 | 7 | 1581 | AAM73503.1 | |
| AY102282.1 | 8 | 1638 | AAM73504.1 | |
| AY102283.1 | 7 | 1141 | AAM73505.1 | |
| AY102284.1 | 9 | 3999 | AAM73506.1 | |
| BC032192.1 | 7 | 2294 | AAH32192.1 |

Genomic Map