Assay Details
Target Gene Details
Entrez Gene ID: | 17762 |
Gene Name: | microtubule-associated protein tau |
Gene Aliases: |
AI413597, AW045860, Mtapt, Tau |
Location: |
Chr.11:104231353-104332090 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapt | NM_001038609.2 | 8 | 918 | NP_001033698.1 |
| NM_001285454.1 | 6 | 744 | NP_001272383.1 | |
| NM_001285455.1 | 5 | 678 | NP_001272384.1 | |
| NM_001285456.1 | 6 | 744 | NP_001272385.1 | |
| NM_010838.4 | 6 | 744 | NP_034968.3 | |
| AF483518.1 | 5 | 559 | AAL90792.1 | |
| AF483519.1 | 5 | 559 | AAL90793.1 | |
| AK078167.1 | 6 | 717 | BAC37157.1 | |
| AK147636.1 | 8 | 879 | BAE28038.1 | |
| BC014748.1 | 6 | 682 | AAH14748.1 | |
| CO432693.1 | ||||
| M18775.1 | 6 | 745 | AAA40165.1 | |
| M18776.1 | 6 | 745 | AAA40166.1 | |
| M93266.1 | 1 | 63 | ||
| U12914.1 | 8 | 802 | AAA58343.1 | |
| U12915.1 | 6 | 628 | AAA58344.1 | |
| U12916.1 | 5 | 562 | AAA58345.1 | |
| Z12133.1 | 1 | 63 | CAA78121.1 |

Genomic Map