Assay Details
Target Gene Details
Entrez Gene ID: | 13447 |
Gene Name: | double C2, beta |
Gene Aliases: |
- |
Location: |
Chr.11:75769090-75796057 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Doc2b | NM_007873.2 | 1 | 180 | NP_031899.2 |
| BC067030.1 | 1 | 180 | AAH67030.1 | |
| D85037.1 | 1 | 172 | BAA12714.1 |

Genomic Map