Assay Details
Target Gene Details
Entrez Gene ID: | 110521 |
Gene Name: | human immunodeficiency virus type I enhancer binding protein 1 |
Gene Aliases: |
Cryabp1 |
Location: |
Chr.13:42051969-42185030 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hivep1 | NM_007772.2 | 8 | 6871 | NP_031798.2 |
| AK134323.1 | 5 | 1263 | BAE22097.1 | |
| X68946.1 | 4 | 631 | CAA48762.1 |

Genomic Map