Assay Details
Target Gene Details
Entrez Gene ID: | 17129 |
Gene Name: | SMAD family member 5 |
Gene Aliases: |
1110051M15Rik, AI451355, Dwf-C, Madh5, MusMLP |
Location: |
Chr.13:56702988-56742379 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smad5 | NM_001164041.1 | 7 | 6141 | NP_001157513.1 |
| NM_001164042.1 | 7 | 5761 | NP_001157514.1 | |
| NM_008541.3 | 7 | 5738 | NP_032567.1 | |
| AK172596.1 | 1 | 846 | ||
| BC048233.1 | 4 | 2005 | AAH48233.1 | |
| BC050001.1 | 7 | 5641 | AAH50001.2 |
More Information
Set Membership: |
|

Genomic Map