Assay Details
Target Gene Details
Entrez Gene ID: | 72308 |
Gene Name: | BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB (S. cerevisiae) |
Gene Aliases: |
2510002F24Rik, GTF3B, TAF3C, TAFIII90, TFIIIB90, mTFIIIB90 |
Location: |
Chr.12:112955631-113000707 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brf1 | NM_028193.3 | NP_082469.2 | ||
| BC037224.1 | AAH37224.1 | |||
| BU148121.1 | ||||
| CF737042.1 |
Target Gene Details
Entrez Gene ID: | 399566 |
Gene Name: | BTB (POZ) domain containing 6 |
Gene Aliases: |
- |
Location: |
Chr.12:112974998-112979493 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btbd6 | NM_001145900.1 | 5 | 1739 | NP_001139372.1 |
| NM_201646.2 | 4 | 1821 | NP_964008.2 | |
| AI118479.1 | 1 | 255 | ||
| AK146991.1 | 4 | 1653 | BAE27593.1 | |
| BC031195.1 | 4 | 1821 | AAH31195.1 |
More Information
Set Membership: |
|

Genomic Map