Assay Details
Target Gene Details
Entrez Gene ID: | 15212 |
Gene Name: | hexosaminidase B |
Gene Aliases: |
- |
Location: |
Chr.13:97176331-97198357 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hexb | NM_010422.2 | 1 | 275 | NP_034552.1 |
| AK151149.1 | 1 | 212 | BAE30155.1 | |
| AK159134.1 | 1 | 228 | BAE34846.1 | |
| AK159178.1 | 1 | 249 | BAE34877.1 | |
| AK160560.1 | 1 | 244 | BAE35874.1 | |
| AK168825.1 | 1 | 249 | BAE40652.1 | |
| AK185127.1 | ||||
| BY145642.1 | 1 | 276 | ||
| U07633.1 | 1 | 247 | AAA18776.1 | |
| Y00964.1 | 1 | 188 | CAA68781.1 |
More Information
Set Membership: |
|

Genomic Map