Assay Details
Target Gene Details
Entrez Gene ID: | 319765 |
Gene Name: | insulin-like growth factor 2 mRNA binding protein 2 |
Gene Aliases: |
C330012H03Rik, IMP-2, Imp2, Neilsen |
Location: |
Chr.16:22059009-22163299 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igf2bp2 | NM_183029.2 | 6 | 956 | NP_898850.2 |
| AK044984.1 | 5 | 472 | ||
| AK049196.1 | 6 | 653 | ||
| AK077404.1 | 6 | 636 | ||
| AK159654.1 | 6 | 689 | ||
| AK170531.1 | 5 | 460 | BAE41861.1 | |
| AY531659.1 | 6 | 620 | AAT01428.1 |
More Information
Set Membership: |
|

Genomic Map