Assay Details
Target Gene Details
Entrez Gene ID: | 13845 |
Gene Name: | Eph receptor B3 |
Gene Aliases: |
AW456895, Cek10, Etk2, HEK2, MDK5, Sek4, Tyro6 |
Location: |
Chr.16:21204795-21223309 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ephb3 | NM_010143.1 | 3 | 752 | NP_034273.1 |
| AK144414.1 | 3 | 768 | ||
| AK158043.1 | 3 | 1813 | BAE34333.1 | |
| BC014822.1 | 3 | 753 | AAH14822.1 | |
| BC053085.1 | 3 | 752 | AAH53085.1 | |
| U11493.1 | 1 | 212 | AAA67925.1 | |
| Z49086.1 | 3 | 610 | CAA88910.1 |
More Information
Set Membership: |
|

Genomic Map