Assay Details
Target Gene Details
Entrez Gene ID: | 64705 |
Gene Name: | dihydropyrimidinase |
Gene Aliases: |
1200017I10Rik, 1300004I01Rik, DHP, DHPase |
Location: |
Chr.15:39768485-39857470 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpys | NM_022722.3 | 10 | 2350 | NP_073559.3 |
| AK004899.2 | 10 | 2347 | BAB23654.1 |
More Information
Set Membership: |
|

Genomic Map