Assay Details
Target Gene Details
Entrez Gene ID: | 100038659 |
Gene Name: | predicted gene 10872 |
Gene Aliases: |
ENSMUSG00000075583, Gm24562 |
Location: |
Chr.15:76266787-76269839 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10872 | NR_045747.1 | 2 | 114 | |
| AK155479.1 | 2 | 115 | BAE33286.1 |
Target Gene Details
Entrez Gene ID: | 74281 |
Gene Name: | spermatogenesis and centriole associated 1 |
Gene Aliases: |
1700084J23Rik, speriolin |
Location: |
Chr.15:76267835-76292577 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spatc1 | NM_028852.1 | 1 | 241 | NP_083128.1 |
| AB032199.1 | 1 | 241 | BAD08243.1 | |
| AK006994.1 | 1 | 242 | BAB24825.1 | |
| BC116725.1 | 1 | 208 | ||
| BC118509.1 | 1 | 201 |

Genomic Map