Assay Details
Target Gene Details
Entrez Gene ID: | 78618 |
Gene Name: | ArfGAP with coiled-coil, ankyrin repeat and PH domains 2 |
Gene Aliases: |
4832442G16, 9530039J15Rik, CNT-B2, Centb2, mKIAA0041 |
Location: |
Chr.16:31092412-31202056 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acap2 | NM_030138.2 | 1 | 254 | NP_084414.1 |
| AB490692.1 | ||||
| AK020591.1 | 1 | 255 | BAB32141.3 | |
| AK029343.1 | 1 | 179 | ||
| AK032916.1 | 1 | 182 | ||
| AK044812.1 | 1 | 245 | BAC32103.1 | |
| AK129041.1 | 1 | 231 | BAC97851.1 | |
| AK147319.1 | 1 | 180 | BAE27843.1 | |
| BB465902.2 | 1 | 297 | ||
| BY014253.1 | 1 | 259 |

Genomic Map