Assay Details
Target Gene Details
Entrez Gene ID: | 239845 |
Gene Name: | G protein-coupled receptor 156 |
Gene Aliases: |
Gababl |
Location: |
Chr.16:37915999-38007530 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr156 | NM_153394.2 | 2 | 497 | NP_700443.2 |
| AF488740.1 | AAN03796.1 | |||
| AK141990.1 | 2 | 541 | ||
| BC059227.1 | 2 | 497 | AAH59227.1 |
More Information
Set Membership: |
|

Genomic Map